Anti-MAT2A Chicken Polyclonal Antibody

Supplier: ProSci
XW-8062
10069-934EA 1269.69 CAD
10069-934
Anti-MAT2A Chicken Polyclonal Antibody
Antibodies
FUNCTION: Catalyzes the formation of S-adenosylmethionine from methionine and ATP.
CATALYTIC ACTIVITY: ATP + L-methionine + H2O = phosphate + diphosphate + S-adenosyl-L-methionine.
SUBUNIT: Homodimer.
SIMILARITY: Belongs to the AdoMet synthetase family.
COFACTOR: Binds 2 divalent ions, such as magnesium or cobalt, per subunit (By similarity).

S-adenosylmethionine synthetase isoform type-2 antibody can be used for the detection of S-adenosylmethionine synthetase isoform type-2 by Western Blot.

Phosphate-Buffered Saline. No preservatives added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MAT2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR