Anti-CEACAM5 Chicken Polyclonal Antibody

Supplier: ProSci
XW-8078
10071-430EA 1269.69 CAD
10071-430
Anti-CEACAM5 Chicken Polyclonal Antibody
Antibodies
PTM: Complex immunoreactive glycoprotein with a MW of 180 kDa comprising 60% carbohydrate.
SIMILARITY: Contains 7 Ig-like (immunoglobulin-like) domains.
SUBCELLULAR LOCATION: Attached to the membrane by a GPI-anchor.
TISSUE SPECIFICITY: Found in adenocarcinomas of endodermally derived digestive system epithelium and fetal colon.

Carcinoembryonic antigen-related cell adhesion molecule 5 antibody can be used for the detection of Carcinoembryonic antigen-related cell adhesion molecule 5 by Western Blot.

Phosphate-Buffered Saline. No preservatives added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CEACAM5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR