Anti-CDKN1B Chicken Polyclonal Antibody

Supplier: ProSci
XW-8094
10069-962EA 1269.69 CAD
10069-962
Anti-CDKN1B Chicken Polyclonal Antibody
Antibodies
Cyclin-dependent kinase inhibitor p27 Kip1 antibody can be used for the detection of Cyclin-dependent kinase inhibitor p27 Kip1 by Western Blot.

Phosphate-Buffered Saline. No preservatives added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CDKN1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR