Anti-CXCL11 Chicken Polyclonal Antibody

XW-8098
10069-966EA 696.47 CAD
10069-966
Anti-CXCL11 Chicken Polyclonal Antibody
FUNCTION: Chemotactive for interleukin-activated T cells but not unstimulated T cells, neutrophils or monocytes. Induces calcium release in activated T cells. Binds to CXCR3. May play an important role in CNS diseases which involve T cell recruitment. May play a role in skin immune responses.
DISEASE: Expressed in a variety of skin disorders, including allergic contact dermatitis, lichen planus and mycosis fungoides.

Small inducible cytokine B11 antibody can be used for the detection of Small inducible cytokine B11 by Western Blot.

Phosphate-Buffered Saline. No preservatives added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CXCL11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR