Anti-CD44 Chicken Polyclonal Antibody

Supplier: ProSci
XW-8104
10070-866EA 1269.69 CAD
10070-866
Anti-CD44 Chicken Polyclonal Antibody
Antibodies
CD44 isoform RC antibody can be used for the detection of CD44 isoform RC by Western Blot.

Phosphate-Buffered Saline. No preservatives added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CD44
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR