Anti-SEC23B Chicken Polyclonal Antibody

Supplier: ProSci
XW-8120
10071-392EA 1269.69 CAD
10071-392
Anti-SEC23B Chicken Polyclonal Antibody
Antibodies
FUNCTION: Component of the COPII coat, that covers ER-derived vesicles involved in transport from the endoplasmic reticulum to the Golgi apparatus. COPII acts in the cytoplasm to promote the transport of secretory, plasma membrane, and vacuolar proteins from the endoplasmic reticulum to the Golgi complex (By similarity).

Protein transport protein Sec23B antibody can be used for the detection of Protein transport protein Sec23B by Western Blot.

Phosphate-Buffered Saline. No preservatives added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SEC23B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR