Anti-TXLNA Chicken Polyclonal Antibody

Supplier: ProSci
XW-8134
10071-778EA 1269.69 CAD
10071-778
Anti-TXLNA Chicken Polyclonal Antibody
Antibodies
FUNCTION: May be involved in intracellular vesicle traffic and potentially in calcium-dependent exocytosis in neuroendocrine cells.
SUBUNIT: Binds to the C-terminal coiled coil region of syntaxin family members STX1A, STX3A and STX4A, but not when these proteins are complexed with SNAP25, VAMP2 or STXBP1, suggesting that it interacts with syntaxins that do not form the SNARE complex.

?-taxilin antibody can be used for the detection of ?-taxilin by Western Blot.

Phosphate-Buffered Saline. No preservatives added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TXLNA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human, Dog, Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR