Anti-TDGF1 Chicken Polyclonal Antibody

XW-8170
10071-768EA 596.37 CAD
10071-768
Anti-TDGF1 Chicken Polyclonal Antibody
FUNCTION: Could play a role in the determination of the epiblastic cells that subsequently give rise to the mesoderm.SIMILARITY: Contains 1 EGF-like domain.
TISSUE SPECIFICITY: Preferentially expressed in gastric and colorectal carcinomas than in their normal counterparts.

Teratocarcinoma-derived growth factor 1 antibody can be used for the detection of Teratocarcinoma-derived growth factor 1 by Western Blot.

Phosphate-Buffered Saline. No preservatives added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TDGF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Bovine, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR