About this item
A slender and handy sensor for simultaneous measurement of conductivity, dissolved oxygen and temperature.
- Immediately ready for use and little maintenance required
- Easily removable oxygen module works according to the galvanic principle
- Fully automatic salinity correction
- Four-electrode system capable of accurate conductivity measurements even when electrodes are dirty
- Choice of cable lengths available
For all water analysis applications in the laboratory and in the field.
Temperature range: 0...+50 °C
Ø×L: 15,3×120 mm