Taq DNA Polymerase, QuantaBio
2200010 2200030 2200020 2200000
10052-246EA 128.22 CAD
10052-246 10052-250 10052-248 10052-244
Taq DNA Polymerase, QuantaBio
  • High yield and specificity
  • Self adjusting Mg2+ buffer technology minimizes PCR optimization
An ultra-pure, thermostable recombinant DNA polymerase offering the highest purity, reproducibility and processivity. The enzyme provides a thermostability that meets the requirements of specialized protocols. An initial incubation of 20 minutes at 95 °C before starting the PCR reaction does not significantly influence the activity of the enzyme.
Product includes: Taq DNA polymerase (5 U/μl), 10X reaction buffer with Mg2+, and separate vial of 25 mM magnesium solution.