PerfectTaq™ DNA Polymerase, QuantaBio
2200070 2200060
10052-258EA 177.05 CAD
10052-258 10052-254
PerfectTaq™ DNA Polymerase, QuantaBio
  • Higher yields compared to other Taq DNA polymerases
  • High sensitivity and specificity
  • Time saving standardised protocols
The PerfectTaq™ DNA polymerase kit provides the components and procedures necessary for efficient and robust standard end point PCR and RT-PCR applications without optimisation.
Product comprises: PerfectTaq™ DNA polymerase (5 U/µl), 10X PCR buffer, 25 mM MgCl₂.