RNase-Free DNase Set, QuantaBio
2500120
10052-210EA 102.32 CAD
10052-210
RNase-Free DNase Set, QuantaBio
  • Efficient on-column and in solution digest of DNA
  • Complete DNase removal by subsequent washes
  • Guaranteed RNase-free
  • Stable, lyophilized enzyme with RNase-Free Buffer and water
The 5 PRIME RNase-Free DNase Set is quality-controlled for DNase Digestion of RNA in Solution before RNA Cleanup or on column DNase Digestion in combination with the PerfectPure RNA Blood Kit. The quantity provided is sufficient for 50 on-column digestions or 200 RNA clean-up reactions. Format, 2500 Kunitz units/mg. One Kunitz unit is defined as the amount of DNase I that causes an increase in A260 of 0.001 per minute per milliliter at 25°C, pH 5.0, with highly polymerized DNA as the substrate.
Ordering information: Kit includes DNase I, RNase-Free (lyophilized; 1500 Kunitz units), 2 x 2 mL DNase Digestion Buffer, and 1.5 mL RNase-Free Water.