QuantaBio MasterMix
2200110 2200100
10052-266EA 771.71 CAD
10052-266 10052-264
QuantaBio MasterMix
  • Ready to use master mix format
  • Minimal optimization with self-adjusting Mg₂+ buffer technology
  • Convenient 4 °C storage eliminates freeze and thaw cycle
For robust amplification with minimal reaction set-up. This 2.5X MasterMix is ready to use and offers high reproducibility for large sample series. Because only primer and template need to be added to the 2.5X concentrate, MasterMix reduces the number of time consuming pipetting steps. This not only reduces the likelihood of errors and the risk of contamination, but also increases precision and sample throughput.
Product includes: Taq DNA polymerase (0.06 U/µl), nucleotides, 25 mM magnesium solution, 2.5X Reaction Buffer and Master Taq to ensure high yields and excellent stability of the reaction components.