TBE 5X, 5PRIME
2500050
10052-202EA 103.92 CAD
10052-202
TBE 5X, 5PRIME
Electrophoresis running buffer for the separation of double-stranded DNA on agarose and polyacrylamide gels
TBE is used as a running buffer for the separation of double-stranded DNA on agarose and polyacrylamide gels. The 5x concentrate is composed of 0.45 M Tris-Borate and 0.01 M EDTA, pH 8.3. It should be diluted to a working concentration of either 1x or 0.5x. The 5- liter size is packaged in an easy dispensing system. Store at room temperature.
DNA samples (specific bands) should resolve under normal gel electrophoretic conditions (agarose and polyacrylamide gels) by using 1x TBE as the final concentration.