THP-1 Lysate, ProSci
Supplier: ProSci
1208
10062-590EA 252.15 CAD
10062-590
THP-1 Lysate, ProSci
Cells
THP-1 lysate was prepared by homogenization in modified RIPA buffer (150 mM sodium chloride, 50 mM Tris-HCl, pH 7.4, 1 mM ethylenediaminetetraacetic acid, 1 mM phenylmethylsulfonyl fluoride, 1% Triton X-100, 1% sodium deoxycholic acid, 0.1% sodium dodecylsulfate, 5 μg/ml of aprotinin, 5 μg/ml of leupeptin
Cell debris was removed by centrifugation. Protein concentration was determined with Bio-Rad protein assay. The THP-1 lysate was boiled for 5 min in 1 x SDS sample buffer (50 mM Tris-HCl pH 6.8, 12.5% glycerol, 1% sodium dodecylsulfate, 0.01% bromophenol blue) containing 5% β-mercaptoethanol.