Recombinant P62 (from E. coli)
Supplier: ProSci
39-906
10072-258EA 883.64 CAD
10072-258
Recombinant P62 (from E. coli)
Proteins and Peptides
Protein p62 preferentially binds multiubiquitin chains and forms a novel cytoplasmic structure "sequestosome" which serves as a storage place for ubiquitinated proteins.
This recombinant protein can be used for WB, ELISA.
20mM Tris-HCl, 150mM NaCl, 1mM EDTA, 5mM Glutathione, 0.5mM GSSH and 10% Glycerol.
Store in working aliquots at -80°C. Avoid freeze/thaw cycles.