Perfect PCR Cloning Kit, QuantaBio
2201310
10052-056EA 399.45 CAD
10052-056
Perfect PCR Cloning Kit, QuantaBio
  • Efficient UA-cloning system with high specificity
  • Simple and quick procedure - approximately 40 minute procedure from cloning to plated cells
For direct cloning of PCR products generated by Taq and other non proofreading DNA polymerases.
Ordering information: Kit contains 2X ligation master mix (200 µl), pPrime cloning vector (2.0 µg), and RNase-free water (1.7 mL)