Plasmid Purification, PerfectPrep Spin Mini Kit, 5PRIME
2300110 2300100
10052-064EA 310.31 CAD
10052-064 10052-062
Plasmid Purification, PerfectPrep Spin Mini Kit, 5PRIME
  • 5 to 20 µg ready to use plasmid DNA in less than 30 minutes
  • Simple bind-wash-elute procedure
  • No resins, slurries or phenol
The PerfectPrep™ Spin Mini kit is a ready to use, complete kit providing all components and procedures necessary for purification of molecular biology grade plasmid DNA. The PerfectPrep™ miniprep system is designed for quick and convenient processing of up to 24 samples simultaneously in less than 30 minutes.
From different amount of starting material up to 20 µg high-quality plasmid DNA can purified using the PerfectPrep Spin Mini. The eluate containing pure supercoiled and open circular plasmid DNA with no other contaminating nucleic acids.