Human Recombinant SCF (from E. coli)
Supplier: ProSci
40-282
10072-078EA 392.66 CAD
10072-078
Human Recombinant SCF (from E. coli)
Proteins and Peptides
Stem Cell Factor (SCF) is a hematopoietic growth factor that exerts its activity by signaling through the c-Kit receptor. SCF and c-Kit are essential for the survival, proliferation and differentiation of hematopoietic cells committed to the melanocyte and germ cell lineages. Human SCF manifests low activity on murine cells, while murine and rat SCF are fully active on human cells. Recombinant human SCF is an 18.4 kDa polypeptide containing 165 amino acid residues, which corresponds to the sequence of the secreted soluble form of SCF.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.