Human Recombinant FLT3 Ligand (from E. coli)
Supplier: ProSci
40-286
10072-872EA 392.66 CAD
10072-872
Human Recombinant FLT3 Ligand (from E. coli)
Proteins and Peptides
Flt3-Ligand is a growth factor that regulates proliferation of early hematopoietic cells. Flt3-Ligand binds to cells expressing the tyrosine kinase receptor Flt3. Flt3-Ligand, by itself does not stimulate proliferation of early hematopoietic cells, but synergizes with other CSFs and interleukins to induce growth and differentiation. Unlike SCF, Flt3-Ligand exerts no activity on mast cells. Multiple isoforms of Flt3-Ligand have been identified. The predominant biologically active form is anchored to the cell surface as the extracellular domain of a transmembrane protein (209 a.a.). The membrane-bound isoform can be proteolytically cleaved to generate a biologically active soluble isoform. Recombinant human Flt3-Ligand is a soluble 17.6 kDa protein consisting of 155 amino acid residues.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.