Human Recombinant Cardiotrophin 1 (from E. coli)
Supplier: ProSci
40-290
10072-876EA 392.66 CAD
10072-876
Human Recombinant Cardiotrophin 1 (from E. coli)
Proteins and Peptides
CT-1 is a member of the IL-6 family of cytokines which also includes LIF, CNTF, OSM (Oncostatin M), IL-11, IL-6 and possibly NT-1/ BSF-3. CT-1 is a pleiotropic cytokine which is expressed in various tissues including the adult heart, skeletal muscle, ovary, colon, prostate and fetal lung and signals through the LIF receptor and the gp130 receptor subunit. CT-1 has the ability to induce cardiac myocyte hypertrophy, and enhances the survival of cardiomyocyte and different neuronal populations. Biologically active human CT-1 is synthesized as a 201 amino acid polypeptide lacking a hydrophobic N-terminal secretion signal sequence. Recombinant Human CT-1 is a 21.5 kDa protein consisting of 201 amino acid residues.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.