Human Recombinant Osteoprotegerin (from E. coli)
Supplier: ProSci
40-323
10072-940EA 392.66 CAD
10072-940
Human Recombinant Osteoprotegerin (from E. coli)
Proteins and Peptides
Osteoprotegerin (OPG) is a member of the TNFR superfamily that can act as a decoy receptor for RANKL. Binding of soluble OPG to sRANKL inhibits osteoclastogenesis by interrupting the signaling between stromal cells and osteoclastic progenitor cells, thereby leading to excess accumulation of bone and cartilage. OPG is expressed in a wide variety of tissues including adult heart, lung, kidney, liver, spleen, prostate, lymph node and bone marrow. OPG is secreted both as a monomeric and a dimeric protein. Its primary structure consists of seven distinct domains, four of which corresponds to the extracellular cysteine-rich domains of TNFR proteins and constitutes the soluble OPG. Recombinant human OPG is a soluble 20.0 kDa protein containing 174 amino acid residues.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.