Human Recombinant Galectin 3 (from E. coli)
Supplier: ProSci
40-329
10072-946EA 392.66 CAD
10072-946
Human Recombinant Galectin 3 (from E. coli)
Proteins and Peptides
Lectins, of either plant or animal origin, are carbohydrate binding proteins that interact with glycoprotein and glycolipids on the surface of animal cells. The Galectins are lectins that recognize and interact with beta-galactoside moieties. Galectin-3 regulates a number of biological processes, including embryogenesis, inflammatory responses, cell progression and metastasis. Galectin-3 is normally expressed in epithelia of a variety of tissues, including colon and endometrium, and in various inflammatory cells, including macrophages. Galectin-3 can function intracellularly, in controlling cell cycle and preventing T-cell apoptosis, and also extracellularly, in activating various cells, including monocytes/macrophages, mast cells, neutrophils, and lymphocytes. Expression of Galectin-3 is affected by neoplastic transformation, being up-regulated in certain types of lymphomas, and in thyroid and hepatic carcinomas. Conversely, it is down-regulated in other cancers, such as colon, breast, ovarian, and uterine. Recombinant human Galectin-3 is a globular 26.0kDa protein containing 250 amino acid residues, but no disulfide bonds.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.