Mouse Recombinant MCP3 (from E. coli)
Supplier: ProSci
40-336
10072-098EA 392.66 CAD
10072-098
Mouse Recombinant MCP3 (from E. coli)
Proteins and Peptides
The MCP proteins belong to the CC chemokine family, and signal through CCR2 and, with the exception of MCP-1, other CCR receptors. The MCP proteins chemoattract and activate monocytes, activated T cells, basophils, NK cells, and immature dendritic cells. The MCP family cross- reacts across species. Recombinant murine MCP-3 is an 8.5 kDa protein containing 74 amino acid residues including the four highly conserved cysteine residues present in the CC chemokines.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.