Order Entry
ContactUsLinkComponent
Human Recombinant APO-Saa (from E. coli)
Human Recombinant APO-Saa (from E. coli)
Catalog #: 10072-648
Supplier:  Prosci
Human Recombinant APO-Saa (from E. coli)
Catalog #: 10072-648
Supplier:  Prosci
Price
$477.84
Log in for availability
Unit of Measure Each (10µG)
Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Gene ID:
    6288
  • Protein/peptide type:
    Recombinant
  • Size:
    0.01 mg
  • Source:
    E. coli
  • Species:
    Human
  • Biological activity:
    For most in vitro applications, apo - SAA exerts its biological activity in the concentration range of 10 - 50 μg/mL.
  • Protein synonyms:
    SAA2
    Serum amyloid A-1 protein
    PIG4
    Amyloid fibril protein AA
    SAA
    TP53I4
  • Protein/peptide name:
    APO-Saa
  • Purity:
    >98%
  • Formulation:
    Lyophilized
  • Cat. no.:
    10072-648

Specifications

About this item

Human apo-SAA is a 104 amino acid polypeptide that circulates primarily in association with high-density lipoproteins (HDL). The level of apo-SAA, normally 1-5 μg/mL in plasma, increases 500-1000 fold within 24 hours of an inflammatory stimulus and, under these conditions, is the most abundant HDL apo-lipoprotein. The human SAA gene codes for a 122 amino acid polypeptide, which contains an 18 amino acid N-terminal signal sequence. Recombinant Apo-SAA is a consensus SAA molecule corresponding to human apo-SAA1alpha except for the presence of an N-terminal methionine and substitution of asparagine for aspartic acid at position 60 and arginine for histidine at position 71(the later two substituted residues are present in apo-SAA2beta).

Store in working aliquots at -20°C. Avoid freeze/thaw cycles.

Customers Who Bought This Also Bought