Human Recombinant BD2 (from E. coli)
Supplier: ProSci
40-131
10072-652EA 392.66 CAD
10072-652
Human Recombinant BD2 (from E. coli)
Proteins and Peptides
Defensins (alpha and beta) are cationic peptides with a broad spectrum of antimicrobial activity that comprise an important arm of the innate immune system. The α-defensins are distinguished from the β-defensins by the pairing of their three disulfide bonds. To date, four human β-defensins have been identified; BD-1, BD-2, BD-3 and BD-4. β-defensins are expressed on some leukocytes and at epithelial surfaces. In addition to their direct antimicrobial activities, they are chemoattractant towards immature dendritic cells and memory T cells. The β-defensin proteins are expressed as the C-terminal portion of precursors and are released by proteolytic cleavage of a signal sequence and, in the case of BD-1 (36 a.a.), a propeptide region. β-defensins contain a six-cysteine motif that forms three intra-molecular disulfide bonds. β-Defensins are 3-5 kDa peptides ranging in size from 33-47 amino acid residues. Recombinant Human BD-2 is a 4.3 kDa protein containing 41 amino acid residues.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.