Mouse Recombinant MIP1G (from E. coli)
Supplier: ProSci
40-340
10072-958EA 392.66 CAD
10072-958
Mouse Recombinant MIP1G (from E. coli)
Proteins and Peptides
MIP-1 gamma is a CC chemokine found in murine blood and a wide variety of murine tissues, with no known human homolog. MIP-1 gamma signals through the CCR1 receptor. MIP-1 gamma chemoattracts neutrophils and also inhibits colony formation of bone marrow myeloid immature progenitors. MIP-1 gamma contains six cysteines including the four highly conserved cysteine residues present in CC chemokines. Recombinant murine MIP-1 gamma is an 11.6 kDa protein containing 101 amino acid residues.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.