Mouse Recombinant Exodus 2 (from E. coli)
Supplier: ProSci
40-341
10072-104EA 392.66 CAD
10072-104
Mouse Recombinant Exodus 2 (from E. coli)
Proteins and Peptides
Exodus-2 is a CC chemokine that can signal through the CCR7 receptor. It is expressed in lymph nodes of certain endothelial cells, and in the spleen and appendix. Exodus-2 chemoattracts T and B lymphocytes and inhibits hematopoiesis. Exodus-2 contains six cysteine residues, including the four conserved cysteines present in CC chemokines. Recombinant murine Exodus-2 is a 12.0 kDa protein containing 110 amino acid residues.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.