Mouse Recombinant BLC (from E. coli)
Supplier: ProSci
40-351
10072-122EA 392.66 CAD
10072-122
Mouse Recombinant BLC (from E. coli)
Proteins and Peptides
BCA-1/BLC, a CXC chemokine, is expressed in the liver, spleen, lymph nodes, appendix and stomach. It exerts its activities through its only receptor CXCR5. BCA-1/BLC is a potent chemoattractant for B lymphocytes and induces weak chemotactic response in T cells and macrophages. It manifests no activity on neutrophils and monocytes. Recombinant murine BLC is a 9.8 kDa protein containing 88 amino acid residues including the four highly conserved cysteine residues present in CXC chemokines.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.