Mouse Recombinant I-Tac (from E. coli)
Supplier: ProSci
40-356
10072-128EA 392.66 CAD
10072-128
Mouse Recombinant I-Tac (from E. coli)
Proteins and Peptides
I-TAC is a "non-ELR" CXC chemokine that is regulated by interferon and signals through the CXCR3 receptor. I-TAC is chemoattractant for IL-2 activated T cells, but does not affect freshly isolated un-stimulated T cells, neutrophils, or monocytes. Recombinant murine I-TAC is a 9.0 kDa protein containing 79 amino acid residues including the four highly conserved cysteine residues present in CXC chemokines.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.