Mouse Recombinant Lungkine (from E. coli)
Supplier: ProSci
40-360
10072-968EA 392.66 CAD
10072-968
Mouse Recombinant Lungkine (from E. coli)
Proteins and Peptides
Lungkine is a CXC chemokine that is expressed in lung epithelial cells and, to a lesser extent, in certain fetal tissues. No human homolog has been identified and a specific cell surface receptor has not yet been found. Lungkine expression in lung tissue is elevated in response to inflammation, at which time it acts to specifically recruit neutrophils and direct them into the lung airway. Recombinant murine Lungkine is a 16.3 kDa CXC chemokine consisting of 142 amino acid residues, including four conserved cysteine residues present in CXC chemokines.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.