RIP1 Peptide, ProSci
Supplier: Prosci
3291P
10062-490EA 197.22 CAD
10062-490
RIP1 Peptide, ProSci
Proteins and Peptides
14 amino acids near the carboxy terminus of human RIP1.
Can be used to block antibody activity completely in WB by incubating with equal volume of antibody for 30 min at 37°C.
PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide