Human BMP15 Peptide
Supplier: ProSci
4739P
10061-754EA 197.22 CAD
10061-754
Human BMP15 Peptide
Proteins and Peptides
18 amino acid peptide near the amino terminus of human BMP15.
Can be used to block antibody activity completely in WB by incubating with equal volume of antibody for 30 min at 37°C.
PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide