Human SIDTR2 Peptide
Supplier: ProSci
7159P
10061-706EA 203.12 CAD
10061-706
Human SIDTR2 Peptide
Proteins and Peptides
18 amino acids near the amino terminus of human SIDTR2.
Can be used to block antibody activity completely in WB by incubating with equal volume of antibody for 30 min at 37°C.
PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide