Human CCNT1 Peptide
Supplier: ProSci
7223P
10061-746EA 203.16 CAD
10061-746
Human CCNT1 Peptide
Proteins and Peptides
18 amino acids near the carboxy terminus of human CCNT1.
Can be used to block antibody activity completely in WB by incubating with equal volume of antibody for 30 min at 37°C.
PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide