Human CD59 Peptide
Supplier: ProSci
7235P
10062-646EA 203.16 CAD
10062-646
Human CD59 Peptide
Proteins and Peptides
14 amino acid peptide near the amino terminus of human CD59.
Can be used to block antibody activity completely in WB by incubating with equal volume of antibody for 30 min at 37°C.
PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide