Human PIBF1 Peptide
Supplier: ProSci
7237P
10062-648EA 203.16 CAD
10062-648
Human PIBF1 Peptide
Proteins and Peptides
15 amino acid peptide near the carboxy terminus of human PIBF1.
Can be used to block antibody activity completely in WB by incubating with equal volume of antibody for 30 min at 37°C.
PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide