Human RC3H1 Peptide
Supplier: ProSci
7251P
10062-658EA 203.16 CAD
10062-658
Human RC3H1 Peptide
Proteins and Peptides
18 amino acid peptide near the carboxy terminus of human RC3H1.
Can be used to block antibody activity completely in WB by incubating with equal volume of antibody for 30 min at 37°C.
PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide