PRKCDBP Peptide, ProSci
Supplier: Prosci
7269P
10062-668EA 203.12 CAD
10062-668
PRKCDBP Peptide, ProSci
Proteins and Peptides
17 amino acid peptide near the center of human PRKCDBP.
Can be used to block antibody activity completely in WB by incubating with equal volume of antibody for 30 min at 37°C.
PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide