Human CNRIP1 Peptide
Supplier: ProSci
7303P
10062-468EA 203.16 CAD
10062-468
Human CNRIP1 Peptide
Proteins and Peptides
15 amino acid peptide near the center of human CNRIP1.
Can be used to block antibody activity completely in WB by incubating with equal volume of antibody for 30 min at 37°C.
PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide