Human UNG2 Peptide
Catalog #: 10062-480
Supplier: Prosci
Price
245,77 $
Log in for availability
Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Specifications
- Size:0.05 mg
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Protein/peptide name:UNG2 Peptide
- Concentration:200 ug/mL
- Formulation:Liquid
- Cat. no.:10062-480
Specifications
About this item
17 amino acid peptide near the amino terminus of human UNG2.
Can be used to block antibody activity completely in WB by incubating with equal volume of antibody for 30 min at 37°C.
PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide
Customers Who Bought This Also Bought