TRIM24 Peptide, ProSci
Supplier: Prosci
7383P
10062-678EA 203.16 CAD
10062-678
TRIM24 Peptide, ProSci
Proteins and Peptides
19 amino acids near the center of human TRIM24.
Can be used to block antibody activity completely in WB by incubating with equal volume of antibody for 30 min at 37°C.
PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide