Rat Recombinant RANTES (from E. coli)
Supplier: ProSci
40-411
10072-388EA 392.66 CAD
10072-388
Rat Recombinant RANTES (from E. coli)
Proteins and Peptides
RANTES is a protein which has been shown to be a chemoattractant for peripheral blood monocytes. It appears to selectively attract T cells of the CD4+/CD45RO+ phenotype in vitro. Recombinant Rat RANTES is a 7.9 kDa protein containing 68 amino acid residues.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.