Human Recombinant FGF19 (from E. coli)
Supplier: ProSci
40-178
10072-720EA 392.66 CAD
10072-720
Human Recombinant FGF19 (from E. coli)
Proteins and Peptides
The FGF family plays central roles during prenatal development and postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF-19, a member of the FGF family, is a high affinity heparin dependent ligand for FGFR4. FGF-19 is expressed during the brain development and embryogenesis. Recombinant human FGF-19 is a 21.8 kDa protein containing 195 amino acid residues.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.