Human NCBP1 Peptide
Supplier: ProSci
7395P
10062-690EA 203.12 CAD
10062-690
Human NCBP1 Peptide
Proteins and Peptides
17 amino acids near the center of human NCBP1.
Can be used to block antibody activity completely in WB by incubating with equal volume of antibody for 30 min at 37°C.
PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide