Heartland virus Heartland Virus Glycoprotein 2 Peptide
Supplier: ProSci
7435P
10062-356EA 203.16 CAD
10062-356
Heartland virus Heartland Virus Glycoprotein 2 Peptide
Proteins and Peptides
19 amino acids near the center of Heartland Virus Glycoprotein 2.
Can be used to block antibody activity completely in WB by incubating with equal volume of antibody for 30 min at 37°C.
PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide