Human Recombinant MIP4 (from E. coli)
Supplier: ProSci
40-385
10072-986EA 392.66 CAD
10072-986
Human Recombinant MIP4 (from E. coli)
Proteins and Peptides
MIP-4 is a CC chemokine that is expressed in lymph nodes, lungs, placenta and bone marrow. MIP-4's primary receptor is unknown. MIP-4 chemoattracts lymphocytes, and has been shown to exert activity on both CD4+ and CD8+ T cells. Recombinant human MIP-4 is a 7.8 kDa protein containing 69 amino acid residues, including the four highly conserved cysteine residues present in CC chemokines.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.