Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Large Capacity Ovens, Forced Air, SHEL LAB

Supplier: Sheldon Manufacturing
SMO28-2
The SHEL LAB forced air ovens feature 27.5 cubic feet of economical oven space

The independent overtemperature safety feature is user adjustable and provides added safety. There are several models to choose from in the SHEL LAB forced air product line, with varying sizes and control features. These large capacity units are perfect for high volume sample and drying applications, including production processes.

The forced, blower-assisted airflow design offers excellent temperature uniformity and fast recovery. A turbo blower and heavy-duty motor combine to direct air over the shelves and samples for even, constant drying, curing and baking. These units are especially ideal for glassware drying. One access port.

Ordering information: Includes 6 shelves (will hold up to 16 max.). 4 internal outlets. Unit does not include a plug and must be hard-wired by an electrician directly into the wall circuit. Two-year limited warranty.



 

ORDER

Expand All / Collapse All
Description Volume Supplier No. VWR Catalog Number Unit Price Quantity
Large Forced Air Oven 781 L (27.5 cu. ft) SMO28-2 89409-466 Each Retrieving
Small Business Enterprise
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Electrical 220V, 5950W, 20A
Exterior dimensions 95.3 x 88.9 x 198.8 cm (37.5 x 35 x 78.3")
Int. WxDxH (mm) 78.1 x 63.5 x 157.4 cm (30.7 x 25 x 62")
Temperature range Ambient +15°C to 260°C
Temperature uniformity +/- 3°C at 110°C
Recovery Time 10 min. at 150°C
Heat-up time 35 min. to 150°C

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty