Allegro II Peristaltic Pump System, KD Scientific
Supplier: KD SCIENTIFIC INC
785502
10117-508EA 7415.91 CAD
10117-508
Allegro II Peristaltic Pump System, KD Scientific
Pumps Peristaltic Pumps
Dual channel, Masterflex® Pump Head, drive and controller
  • Allows maintenance free, continuous flow pumping of various liquids
  • Clearly displays flow rate and total dispensed volume, adjustable flow rate while pump is running
  • User configurable multi-step operation
  • Dispenses in forward or reverse direction
  • Stackable sealed remote pump drive w/ ip 32 rating
  • Selectable tubing table
  • External connectivity with USB interface and I/O port
  • Two simultaneous individual flows or one combined flow
Provides flow rates up to 720 ml/min (tubing dependant) using combined channels. Accepts tubing ID sizes 0.8 mm (0.03") - 8.0 mm (0.32 in). Compatible with Masterflex® L/S® 13, L/S® 14, L/S® 16, L/S® 25 tubing with durometer range 55A-62A.
Certifications: UL, CSA, CUL, CE (Global), RoHS, WEEE.