Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • MWBE (4) SBE (4)
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Tape Dispensers

https://ca.vwr.com/stibo/smallweb/std.lang.all/96/76/18709676.jpg

Less expensive and bulky than automated machines, tape dispensers are equally as productive for heavy every day use. Capable of one-handed operation, adhesive taping duties are easy. Choose between specific millimeter core size dispensers or an adjustable core. Multi-roll accommodating styles include separating discs to keep rolls from sticking. For essential identification, tape dispensers with a large writing surface facilitate clearer sample labeling. Both steel and plastic serrated edges cut tape evenly and quickly.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"16"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Catalog Number: (89098-146)
Supplier:  VWR International
Description:   This durable plastic handheld dispenser for tapes and labels accommodates a maximum width of 2.9 cm (11/8").
Catalog Number: (89098-148)
Supplier:  VWR International
Description:   Single-roll dispenser is designed with an adjustable core size to hold either a 12.7m (41.7') long roll of tape on a 25.4 mm (1") core or a 54.9m (180...
Supplier:  VWR International
Description:   Tape dispensers are constructed of steel for extra protection.
Supplier:  Bel-Art Products
Description:   Write-On™ Label Tape Dispensers provide a smooth writing surface, making labeling easy and fast.
Supplier:  Bel-Art Products
Description:   Convenient dispenser holds and releases tape cut to user specifications.
Supplier:  Micronova
Description:   Designed for use with Micronova tapes
Minority or Woman-Owned Business Enterprise Small Business Enterprise
Supplier:  Bel-Art Products
Description:   Scienceware® Write-On™ Labeling Tape or any tape with a 7.6 cm (3") ID core will fit on the drum of this multi-roll dispenser, which includes disks to...
Supplier:  Bel-Art Products
Description:   The protective labeling system provides labels that stand up to harsh laboratory conditions.
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 16
  1  2  Next